Aniline blue, OR, certified by the Biological Stain Commission, Macron Fine Chemicals™

Synonyms: Acid Blue 22Marine Blue VDisodium hydrogen aminomethyl[[4-[(sulphonatophenyl)amino]phenyl][4-[(sulphonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulphonateChina BlueCotton BlueCI 42755Blackley BlueCN DcK-11Water Blue

Management of Change (MOC) category = NI

H179-55 MACRH179-55
MKH17955EA 98.28 USD
CAMKH17955 MKH17955
Aniline blue, OR, certified by the Biological Stain Commission, Macron Fine Chemicals™
Aniline blue

Aniline Blue/WS, OR, Certified by Biological Stain Commission

Formula: C₃₂H₂₅N₃Na₂O₉S₃
MW: 737.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036130
CAS Number: 28631-66-5
EINECS: 249-113-9

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Total Dye Content (mL/0.1N TiCl₃/gm), For Information Only
Absorbance Maximum, nm, For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR