Ammonium fluoride 40% in aqueous solution, Finyte®, J.T.Baker®

Danger


Management of Change (MOC) category = R

5864-09 BAKR5864-09
JT5864-9EA 2851.02 USD
JT5864-9 CAJT5864-9
Ammonium fluoride 40% in aqueous solution, Finyte®, J.T.Baker®
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

For Microelectronic Use
Assay (NH₄F) 40.0 - 41.0 %
Color (APHA) ≤10
pH of 1% Solution at 25°C 6.0 - 7.5
Chloride (Cl) ≤2 ppm
Nitrate (NO₃) ≤3 ppm
Phosphate (PO₄) ≤0.20 ppm
Sulfate (SO₄) ≤1 ppm
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Trace Impurities - Antimony (Sb) ≤5.0 ppb
Trace Impurities - Arsenic (As) ≤5.0 ppb
Arsenic and Antimony (as As) ≤10.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤10.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Trace Impurities - Iron (Fe) ≤25.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count at point of fill - 0.5 µm and greater
Particle Count at point of fill - 1.0 µm and greater ≤10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR