Nickel (II) sulfate hexahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nickelous sulphate, HexahydrateNickelous sulfate, HexahydrateNickel sulphate hexahydrateNickel sulfate hexahydrate

Management of Change (MOC) category = N

BAKR2808-07 2808-07 2808-01 BAKR2808-04 BAKR2808-01
JT2808-7EA 4384.44 USD
JT2808-7 CAJT2808-1 CAJT2808-7 JT2808-1 JT2808-4
Nickel (II) sulfate hexahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Nickel sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3077
ADR: 9,III
Merck Index: 13,06541

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NiSO₄·6H₂O) (by EDTA titrn) 98.0 - 102.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Nitrogen Compounds (as N) ≤0.002 %
Calcium (Ca) ≤0.005 %
Cobalt (Co) ≤0.002 %
Copper (Cu) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.005 %
Manganese (Mn) ≤0.002 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR