Ammonium fluoride ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Danger


Management of Change (MOC) category = N

BAKR0698-07 BAKR0698-01 BAKR0698-05
JT0698-7EA 3803.62 USD
JT0698-7 JT0698-5 JT0698-1
Ammonium fluoride ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160 °C
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 12,00553

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NH₄F) ≥98.0 %
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.010 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR