Sodium nitrite USP, Ph. Eur., Extra Pure, SAFC®
|
Danger
|
1.06544.1000
1.06544.5000
1.06544.1000
1.06544.9050
1.06544.5000
1.06544.9050
EM1.06544.5000CS
2278.83
USD
CA1.06544.1000
EM1.06544.5000
CA1.06544.9050
EM1.06544.1000
CA1.06544.5000
EM1.06544.9050
Sodium nitrite USP, Ph. Eur., Extra Pure, SAFC®
Sodium nitrite
Formula:
NaNO₂ MW: 69 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 284 °C Density: 2.168 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 EINECS: 231-555-9 UN: 1500 ADR: 5.1 (6.1),III Merck Index: 12,08793 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...