JACKSON SAFETY® R30 Particulate N99 Respirator with Valve, KIMBERLY-CLARK PROFESSIONAL®
64520 64520
94024-092CS 300.75 USD
94024-092 CA94024-092
JACKSON SAFETY® R30 Particulate N99 Respirator with Valve, KIMBERLY-CLARK PROFESSIONAL®
The R30 Particulate N99 Respirator with Valve delivers an optimal combination of protection and comfort
Durable, woven, ultrasonically bonded elastic head straps help keep respirators securely in place without pulling or breaking. The nose pad features soft foam for added comfort. Full perimeter foam and highly visible, adjustable red head strap are included. The exhalation valve may be used to release uncomfortable moist air. Designed to provide protection against dry and oily particulates, respirators are best suite for use in grinding, sanding, sweeping, woodworking, and machining applications. Latex-free. Packaged in 8 boxes of 10 per box.
Certifications: NIOSH approved.