JACKSON SAFETY® R20 Particulate P95 Respirator With Valve, KIMBERLY-CLARK PROFESSIONAL®
64420 64420
94024-090CS 239.99 USD
94024-090 CA94024-090
JACKSON SAFETY® R20 Particulate P95 Respirator With Valve, KIMBERLY-CLARK PROFESSIONAL®
The R20 Particulate P95 Respirator with Valve delivers an optimal combination of protection and comfort
NIOSH approved.
Durable, woven, ultrasonically bonded elastic head straps help keep respirators securely in place without pulling or breaking. The nose pad features soft foam for added comfort. Highly visible, woven yellow head straps are included. The exhalation valve may be used to release uncomfortable moist air. Designed to provide protection against dry and oily particulates, respirators are best suite for use in grinding, sanding, sweeping, woodworking, and machining applications. Latex-free. Packaged in box. 10 Masks per box, 8 Boxes per case.