JACKSON SAFETY® R10 Particulate N95 Respirator with Comfort Straps and Dual Valves, KIMBERLY-CLARK PROFESSIONAL®
64260 64260
89131-954CS 186.32 USD
89131-954 CA89131-954
JACKSON SAFETY® R10 Particulate N95 Respirator with Comfort Straps and Dual Valves, KIMBERLY-CLARK PROFESSIONAL®
Innovative new respirator design provides comfort, protection, and style
NIOSH approved.
The JACKSON SAFETY® R10 Particulate Respirator with Comfort Straps and Dual Valves represents a major advance in protective comfort. It features an innovative, new head strap design that's preferred over the headstraps of leading N95 valved respirators. Ideally suited for grinding, sanding, sweeping, and woodworking applications. Respirator features wide, comfortable, and adjustable head straps. Dual exhalation valves are positioned to help reduce eyewear fogging. A soft nose foam pad with a cloth layer offers increased comfort throughout extended use. Latex-free. NIOSH N95 approved.