Lithium chloride ≥99%, GR ACS, Supelco®

Warning

LX0331-1 LX0331-3 LX0331-1 LX0331-3
EM-LX0331-3EA 1170.88 USD
CALX0331-3 EM-LX0331-3 EM-LX0331-1 CALX0331-1
Lithium chloride ≥99%, GR ACS, Supelco®
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Assay (argentometric) 99% min
Barium (Ba) 0.003% max
Calcium (Ca) 0.01% max
Color White to off-white
Form Powder or fine granules
Heavy metals (as Pb) 0.002% max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Loss on drying (105°C) 1.0% max
Potassium (K) 0.01% max
Sodium (Na) 0.20% max
Sulfate (SO4) 0.01% max
Titrable base 0.008meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR