ICNA51001
0151001.5 015100101 0151001H05 0151001H.5 015100105 0151001H01
IC51001H01EA 1054.52 USD
IC51001H01 IC5100105 IC51001.5 IC5100101 IC51001H.5 IC51001H05
ICNA51001
Sp.Act. 1000 Ci/mmol; 37 TBq/mmol Solution in 50 mM L-lysine pH 7.4, containing 10 mM 2-mercaptoethanol Concentration ~10 mCi/mL; ~370 MBq/mL Packaged under nitrogen in the Versatainer™ System Shipped in dry ice