Deoxycytidine-5′-Triphosphate, [α-32P] IsoBlue™ Stabilized, MP Biomedicals
0139011X01 0139011X.2 0139011X.5
IC39011X.2EA 479.07 USD
IC39011X.2 IC39011X.5 IC39011X01
Deoxycytidine-5′-Triphosphate, [α-32P] IsoBlue™ Stabilized, MP Biomedicals
(dCTP α-32P) Tetra-triethylammonium Salt Sp
Act. 3000 Ci/mmol; 111 TBq/mmol Stabilized aqueous solution 10 mCi/mL; 370 MBq/mL