Adenosine 5′-Triphosphate, [γ-32P], MP Biomedicals
013502005 013502010
IC3502005EA 605.16 USD
IC3502005 IC3502010
Adenosine 5′-Triphosphate, [γ-32P], MP Biomedicals
Adenosine 5'-Triphosphate, [γ-32P] (ATP γ-32P)
  • End-Labeling Grade
  • Sp. Act. 7,000 Ci/mmol; 259 TBq/mmol
  • 90% ATP [-32P]
  • Aqueous solution, 100 mCi/ml; 3.> GBq/ml
Ref.: Carpenter, C.D. and Simon, A.E., "Simplified RNA sequencing using dideoxy chain termination technique.", Biotechniques, 8: 26-27 (1990).