Adenosine 5′-Triphosphate, [γ-32P], MP Biomedicals
013500101 0135001.5 0135001.2
IC35001.5EA 762.49 USD
IC35001.5 IC35001.2 IC3500101
Adenosine 5′-Triphosphate, [γ-32P], MP Biomedicals
(ATP γ-32P) Tetra-triethylammonium Salt Sp
Act. 25 Ci/mmol; 925 GBq/mmol 5 mM Tris HCl, pH 7.4-7.5 10 mCi/mL; 370 MBq/mL Shipped in dry ice.