Deoxycytidine-5′-Triphosphate, [α-32P], MP Biomedicals
0133004X.5 0133004X.2 0133004X01
IC33004X01EA 1140.69 USD
IC33004X01 IC33004X.2 IC33004X.5
Deoxycytidine-5′-Triphosphate, [α-32P], MP Biomedicals
(dCTP α-32P) Tetra-triethylammonium Salt Sp
Act. 3000 Ci/mmol; 111 TBq/mmol 5 mM Tris HCl, pH 7.4-7.5 10 mCi/mL; 370 MBq/mL Shipped in dry ice.