D-Glucosamine, [6-3H], MP Biomedicals
0127021S.2 0127021S01 0127021S05
IC27021S05EA 2521.54 USD
IC27021S05 IC27021S01 IC27021S.2
D-Glucosamine, [6-3H], MP Biomedicals
Hydrochloride Sp
Act. 25-40 Ci/mmol; 0.925-1.48 TBq/mmol Sterile aqueous solution HPLC Analyzed for Purity