Uridine, [5-3H], MP Biomedicals
0124048S05 0124048S.2 0124048S01
IC24048S01EA 766.76 USD
IC24048S01 IC24048S.2 IC24048S05
Uridine, [5-3H], MP Biomedicals
Sp
Act. 15-30 Ci/mmol; 0.55-1.11 TBq/mmol Sterile aqueous solution HPLC Analyzed for Purity