ICNA20039
0120039H.2 0120039H05 0120039H01
IC20039H01EA 501.6 USD
IC20039H01 IC20039H.2 IC20039H05
ICNA20039
Sp. Act. 5-15 Ci/mmol; 185-555 GBq/mmol Purity 94% Ethanol: water solution (3:7). Containing 10 mM 2-mercaptoethanol Shipped in dry ice.