Formic Acid, [14C], MP Biomedicals
011209191 011209150
IC1209191EA 1138.11 USD
IC1209191 IC1209150
Formic Acid, [14C], MP Biomedicals
Sodium Salt Sp
Act. 40-60 mCi/mmol; 1.48-2.22 GBq/mmol Ethanol: water solution (7:3). HPLC Analyzed for Purity