D-Fructose, [U-14C], MP Biomedicals
011102891 011102883
IC1102891EA 2176.49 USD
IC1102891 IC1102883
D-Fructose, [U-14C], MP Biomedicals
Sp.Act
200-300 mCi/mmol; 7.4-11.1 GBq/mmol Ethanol: water solution (2:8) HPLC Analyzed for Purity