Would you like to log in again?
PRC Prepacked Chromatography Columns, Pall Laboratory
Supplier: Pall Laboratory
PRC08X100HEA PRC05X050CMC
PRC Prepacked Chromatography Columns are ready-to use products for laboratory and process development applications.
  • Prepacked
  • Connect directly to chromatography systems
  • Scalable
They allow users to screen Pall ion exchange and mixed-mode chromatography sorbents under reliable and reproducible conditions
The PRC Prepacked Chromatography Columns are scalable. Two or more of these versatile chromatography columns can be connected in series to increase the column height and more closely model real pilot-scale conditions or scale-down applications. Performance is guaranteed run after run.
They connect directly to commonly-used laboratory chromatography systems, such as AKTA systems.
Ordering information: PRC Prepacked Chromatography Columns are available in 1 mL and 5 mL volumes. Connections are built-in 10-32 fittings. Body and end caps of columns are made of molded polypropylene. Their 17 µm frit is made of PP/PE (polypropylene/polyethylene). HyperCel sorbents are stored in 30% isopropanol/100 mM sodium phosphate, pH 4.3. Ceramic HyperD F sorbents are stored in 20% ethanol/150 mM NaCl.
ORDER
Expand All / Collapse All
Description Column Format Capacity I.D. x L Packing Material Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
PRC Column 5x50 CM Ceramic HyperD F Analytical 1 mL 5 x 50 mm Carboxymethyl 50 µm PRC05X050CMC 89202-170 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PRC Column 8x100 HEA HyperCel Analytical 5 mL 8 x 100 mm Hexylamine (aliphatic) (pKa = 8.0) 90 µm PRC08X100HEA 89202-180 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Description 89202-180 89202-170
Packing Material Mixed-mode Ion exchange
Fittings 10-32 fittings
Max. Pressure HyperCel sorbent: 0.5 barg (7 psig); Ceramic HyperD F sorbent: 1.5 barg (22 psig); 1 mL columns: 20 barg (290 psig); 5 mL columns: 30 barg (435 psig)
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty