    Sort by:
  • Search Within Results
5-Chloro-2'-deoxyuridine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   5-Chloro-2'-deoxyuridine (CldU) is a thymidine analog that is readily incorporated, following phosphorylation, into newly synthesized DNA in place of ...