You Searched For: Undecanenitrile
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCU0044-025G)
Supplier: TCI America
Description: CAS Number: 2244-07-7 MDL Number: MFCD00001945 Molecular Formula: C11H21N Molecular Weight: 167.30 Purity/Analysis Method: 95.0% (GC) Form: C...