    Sort by:
  • Search Within Results
Dodecanedinitrile
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Catalog Number: (AAAL03451-06)
Supplier:  Alfa Aesar
Description:   1,10-Dicyanodecane Dodecanedinitrile. Grade: 98, Melting Point C16-19*. Boiling Point C: 195*/8mm. C12H20N2. 4543-66-2.