    Sort by:
  • Search Within Results
nucleotides
https://us.vwr.com/stibo/smallweb/std.lang.all/95/99/18989599.jpg
Labeled nucleotides come in different concentrations and specific activities to meet any laboratory procedure need. These organic molecules are commonly used to detect specific nucleic acid sequences. The nucleotides may be incorporated in a variety of methods including in vitro transcription with RNA polymerase, 3’ end labeling with terminal deoxynucleotidyl transferase, DNA polymerase, random primed DNA labeling with Klenow fragment, and PCR labeling with thermophilic DNA polymerases like Taq or Pfu.
SearchPresentationType-VERTICAL
SearchResultCount:"34"
Sort by:
Supplier:  MP Biomedicals
Description:   Reliable performance and consistent quality.
Supplier:  MP Biomedicals
Description:   Reliable performance and consistent quality.
Supplier:  MP Biomedicals
Description:   Reliable performance and consistent quality.
Supplier:  MP Biomedicals
Description:   Radiolabeled Nucleotides
Supplier:  MP Biomedicals
Description:   Radiolabeled Nucleotides
Supplier:  MP Biomedicals
Description:   Sp.Act. 60-90 Ci/mmol; 2.22-3.33 TBq/mmol Sterile aqueous solution. HPLC Analyzed for Purity
Supplier:  MP Biomedicals
Description:   (ATP γ-32P) Tetra-triethylammonium Salt Sp. Act. 25 Ci/mmol; 925 GBq/mmol 5 mM Tris HCl, pH 7.4-7.5 10 mCi/mL; 370 MBq/mL Shipped in dry ice.
Supplier:  MP Biomedicals
Description:   Sp.Act. 40-60 Ci/mmol; 1.48-2.22 TBq/mmol Ethanol: water solution (7:3). HPLC Analyzed for Purity
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 34
  1  2  3  4  5  Next