Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® 3L Disposable Spinner Flask, with MPC Accessory

Supplier: Corning
3559
The Corning 3L spinner flask body is constructed of polystyrene, paddle and shaft from polypropylene and caps from high density polyethylene

All are made of USP Class VI and ISO 10993-18 compliant materials. Recommended working range of 1L to 3L. Extended paddle design ensures constant top-to-bottom motion of the liquid without having to adjust the height of the paddle.

Bottom contours on the paddle hug the shape of the flask, eliminating dead spots. Two wide neck openings for easy pouring; easily accept a 100 mL pipet. Vertical side baffles break up laminar flow - ensures better mixing and maximizes cell production. Each system is supplied individually double bagged. Sterilized by gamma irradiation; have the highest Sterility Assurance Level (SAL) of 10-6 and meets USP XXIV:71 Bacteriostasis, Fungistasis and Sterility Standards.

Accessories information: Optional Vented Caps (Cat. No. 89089-914) and Aseptic Transfer Caps (Cat. No. 89089-910) are available.

Ordering information: Single use only. Flasks cannot be autoclaved or resterilized with alcohol. Any attempt to do so will result in degradation of the materials.



 

ORDER

Expand All / Collapse All
Description Material Working Volume Sterility Supplier No. VWR Catalog Number Unit Price Quantity
Corning® 3L Disposable Spinner Flask, Solid Cap, Sterile, with MPC Accessory Plastic 3 L (Max.) Sterile 3559 89214-354 Case of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Thread Finish 45 GL
Cap Ø 54 mm (approx.)
Cap Material High Density Polyethylene

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty