Corning® 3L Disposable Spinner Flask
Supplier: Corning
3579
89166-458CS 3068.06 USD
89166-458
Corning® 3L Disposable Spinner Flask
Flasks Spinner Flasks
The Corning 3L spinner flask body is constructed of polystyrene, paddle and shaft from polypropylene and caps from high density polyethylene
Aseptic Transfer Caps are sterile (10-6 SAL), individually double bagged and fit the side arms on 1L Corning® Disposable Spinner Flasks (Cat. No. 89089-804). Cap is made of high density polyethylene; autoclaving is not recommended. The venting port ends in a 25 mm diameter vent filter with a 0.2 µm pore PVDF membrane and a barbed fitting. The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a 1/8 inch ID (3.2 mm), 1/4 inch OD chemically resistant, heat sealable flexible tubing, ending in a male luer lock (MLL) connector closed with a female luer sealing cap. Style_NA value represents the Flask Style.
All are made of USP Class VI and ISO 10993-18 compliant materials