Would you like to log in again?
VWR® Stainless Steel Chest Freezer/LN2 Racks for 96-/384-Well Microtiter Plates
Supplier: VWR International
Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.
  • Stainless steel construction
  • Durable
  • Sturdy handles
Racks feature sturdy handles. Racks accommodate 96/384 well microtiter plates and contain locking rods. Reorder by simply scanning the QR code with your smartphone or tablet.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Capacity For Use With Dimensions VWR Catalog Number Unit Price Quantity
Stainless Steel Rack for Chest Freezer/LN2, 96-/384-Well Microtiter Plates with Locking Rod 25 Plates with Lid, 30 Plates without Lid, 5 Shelves High 96-/384-Well Microtiter Plates 421H x 92W x 140L mm (169/16 x 35/8 x 51/2)" 89214-604 Each Retrieving
Small Business Enterprise
Stainless Steel Rack for Chest Freezer/LN2, 96-/384-Well Microtiter Plates with Locking Rod 40 Plates with Lid, 48 Plates without Lid, 8 Shelves High 96-/384-Well Microtiter Plates 673H x 92W x 140L mm (261/2 x 35/8 x 51/2)" 89214-608 Each Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty