Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Screw Neck Diagnostic Bottles, Wheaton

Supplier: DWK Life Sciences
240676-05 240676-04 240676-03 240676-02 219371 240676-01
Wheaton Diagnostic Bottles are an alternative to serum bottles and vials making them functional for lyophilization applications

Diagnostic bottles can be autoclaved.

Wheaton I-Loc Closures are designed for use with Wheaton diagnostic bottles. It is a polypropylene screw cap with a gray bromobutyl/50 stopper. The contents of the bottle can be accessed using a syringe through the stopper or with a pipette after removing the screw cap. I-Loc closures can be autoclaved.

Each bottle has a controlled ID which allows for fit with the Wheaton I-Loc Closure or with a 20-400 Screw Cap with Thin Flange Stopper. The product is made from borosilicate glass that conforms to USP Type I requirements.



 

ORDER

Expand All / Collapse All
Capacity Dia. x H Cap Size Color Supplier No. VWR Catalog Number Unit Price Quantity
10 mL 25 x 61 mm 20-400 Amber 219371 15507-118 Case of 480 Retrieving
Small Business Enterprise

Screw Caps
Description Supplier No. VWR Catalog Number Unit Price Quantity
Black I-Loc Closure, Cap size 20-400 240676-01 16149-151 Case of 100 Retrieving
Small Business Enterprise
Blue I-Loc Closure, Cap size 20-400 240676-04 16149-157 Case of 100 Retrieving
Small Business Enterprise
Red I-Loc Closure, Cap size 20-400 240676-03 16149-155 Case of 100 Retrieving
Small Business Enterprise
White I-Loc Closure, Cap size 20-400 240676-02 16149-153 Case of 100 Retrieving
Small Business Enterprise
Yellow I-Loc Closure, Cap size 20-400 240676-05 16149-159 Case of 100 Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty