CiDecon® ST, Sterile Concentrated Disinfectant, Decon Labs
8502
89233-186CS 282.66 USD
89233-186
CiDecon® ST, Sterile Concentrated Disinfectant, Decon Labs
CiDecon® ST Sterile concentrated phenolic disinfectant is individually double-bagged, gamma-irradiated,and tested to ensure sterility
Diluted at a ratio of one ounce per gallon (1:128), CiDecon® ST provides broadspectrum efficacy against a wide variety of organisms, including TB, in the cleanroom environment. 2 oz. SinglServ™ is pre-measured to make 2 gallons of working solution at the correct dilution. Conforms to OSHA’s Bloodborne Pathogens Standard. Each case is shipped with a Lot Specific Document detailing parameters of QC, irradiation, and sterility. EPA registered.