SHETIB01140

IB01140
87004-974EA 54.22 USD
87004-974
SHETIB01140
NP-40 is a non-ionic detergent used to denature cell membranes without denaturing the protein.

  • Appearance: Colorless, viscous liquid
  • Refractive index: 1.48-1.52

Dnase-, Rnase-, and Protease- Free. Store at room temperature, protect from moisture. May become turbid after storage then heat to 40°C to clear product.

NP-40 detergent is used to solubilize cerbral GABA receptors and solubilization of proteins and lipids.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR