Corning® 1L Disposable Spinner Flask
Supplier: Corning
3546
89214-352CS 2522.48 USD InStock
89214-352
Corning® 1L Disposable Spinner Flask
Flasks Spinner Flasks
The Corning 1L spinner flask body is constructed of polystyrene, paddle and shaft from polypropylene and caps from high density polyethylene
All are made of USP Class VI and ISO 10993-18 compliant materials. Recommended working range of 300 mL to 1L. Extended paddle design ensures constant top-to-bottom motion of the liquid without having to adjust the height of the paddle.
Bottom contours on the paddle hug the shape of the flask, eliminating dead spots. Two wide neck openings for easy pouring; easily accept a 100 mL pipet. Vertical side baffles break up laminar flow - ensures better mixing and maximizes cell production. Sterilized by gamma irradiation; have the highest Sterility Assurance Level (SAL) of 10-6 and meets USP XXIV:71 Bacteriostasis, Fungistasis and Sterility Standards.
Accessories information: Optional Vented Caps (Cat. No 89089-914) and Aseptic Transfer Caps (Cat. No. 89089-908) are available.
Ordering information: Single use only. Flasks cannot be autoclaved or resterilized with alcohol. Any attempt to do so will result in degradation of the materials.
Packaging: Each system is supplied individually double bagged.