SHET05040
05040
IB05040EA 51.81 USD
IB05040
SHET05040
Chloroform is a primary component used in nucleic acid, DNA, and RNA extraction procedures to aid in the complete removal of phenol from the aqueous sample.
Store at room temperature and keep tightly sealed. Dnase, Rnase, and Protease Free.