Would you like to log in again?
96-Well Total RNA Kit, IBI Scientific
Supplier: IBI Scientific
IB47360 IB47361
The 96-Well Total RNA Kit is specially designed for high-throughput purification of total RNA from up to 5x10^6 animal cultured cells
Note: For research purposes only.
Using either vacuum or centrifuge, RNA preparations of 96/192 samples can be completed in approximately one hour. This method uses chaotropic salt to lyse cells and inactivate RNase. The RNA in the chaotropic salt is bound by the glass fiber matrix of each well. Once the contaminants have been removed, the purified RNA is eluted by RNase-Free water. The purified RNA is now ready to use in various downstream applications. The sample size is 5x10^6 animal cultured cells.
ORDER
Expand All / Collapse All
Description Binding Capacity Expectant Yield Kit Size Supplier No. VWR Catalog Number Unit Price Quantity
96-Well Total RNA Kit 30 µg 5-10µg for high-copy plasmids; 0.5-5µg for low-copy plasmids 10 x 96-Well IB47361 95043-964 Pack of 10 Retrieving
96-Well Total RNA Kit 30 µg 5-10µg for high-copy plasmids; 0.5-5µg for low-copy plasmids 4 x 96 Preps IB47360 95043-962 Pack of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Binding Capacity Up to 30µg per well
Operation Time 40 minutes
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty