Would you like to log in again?
H-Series ISFET pH Electrodes, Hach
Supplier: Hach
PHW47-SS PH17-SS PHW57-SS PH77-SS PH37-SS PHW37-SS PH57-SS PH47-SS PHW17-SS PHW77-SS
The stainless steel, non-glass ISFET pH probe is ideal for most applications
ISFET technology is excellent for field work, food testing, soil testing, industrial applications, and educational use. Store dry.
Choose a probe that best fits your pH testing needs. Specific connector for benchtop meters.
ORDER
Expand All / Collapse All
Description For Use With pH Range Supplier No. VWR Catalog Number Unit Price Quantity
General-Purpose Round Tip Stainless Steel pH Probe H150/260/270/280 0–14 PH77-SS 82007-534 Each Retrieving Restricted
Product available on GSA Advantage®
General-Purpose Round Tip Stainless Steel pH Probe with Waterproof Connector H160/170/180 0–14 PHW77-SS 82026-950 Each Retrieving Restricted
Product available on GSA Advantage®
Heavy-Duty Meat Handle Stainless Steel Micro pH Probe H150/260/270/280 0–14 PH57-SS 82007-538 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel Heavy-Duty Meat Handle Piercing Tip Micro pH Probe with Waterproof Connector H160/170/180 0–14 PHW57-SS 82026-958 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel Micro pH Probe H150/260/270/280 0–14 PH17-SS 14221-752 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel Micro pH Probe with Waterproof Connector H160/170/180 0–14 PHW17-SS 82026-952 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel pH Probe for NMR Tubes H150/260/270/280 0–14 PH47-SS 14221-756 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel pH Probe for NMR Tubes with Waterproof Connector H160/170/180 0–14 PHW47-SS 82026-956 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel Piercing Tip Micro pH Probe H150/260/270/280 0–14 PH37-SS 14221-754 Each Retrieving Restricted
Product available on GSA Advantage®
Stainless Steel Piercing Tip Micro pH Probe with Waterproof Connector H160/170/180 0–14 PHW37-SS 82026-954 Each Retrieving Restricted
Product available on GSA Advantage®
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty