Methyl-3-methoxy-2-methylbenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-methoxy-2-methylbenzoate
070799-1G
070799-5G
070799-500MG
101946-376EA
150.77
USD
101946-376
101946-378
101946-380
Methyl-3-methoxy-2-methylbenzoate ≥95%
Methyl-3-methoxy-2-methylbenzoate
Hazard: Irritant
Formula:
C₁₀H₁₂O₃ MW: 180.21 g/mol |
MDL Number:
MFCD11846427 CAS Number: 42981-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...