1-Oxa-5-azaspiro[2.3]hexane-5-carboxylicacid phenylmethyl ester ≥95%
Supplier: Matrix Scientific
070750-100MG
070750-250MG
070750-500MG
101946-196EA
1396.92
USD
101946-196
101946-200
101946-198
1-Oxa-5-azaspiro[2.3]hexane-5-carboxylicacid phenylmethyl ester ≥95%
1-Oxa-5-azaspiro[2.3]hexane-5-carboxylicacid phenylmethyl ester
Hazard: Irritant
Formula:
C₁₂H₁₃NO₃ |
MDL Number:
MFCD19443903 CAS Number: 934664-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...