2-Fluoro-3-iodopyrazine ≥95%
Supplier: Matrix Scientific
070748-5G
070748-1G
101946-186EA
815.38
USD
101946-186
101946-188
2-Fluoro-3-iodopyrazine ≥95%
2-Fluoro-3-iodopyrazine
Hazard: Irritant
Formula:
C₄H₂FIN₂ |
MDL Number:
MFCD17677156 CAS Number: 206278-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...