Ethyl-2-(1,3-benzothiazol-2-yl)-2-oxoacetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(benzo[d]thiazol-2-yl)-2-oxoacetate
070742-5G
070742-1G
101946-152EA
884.62
USD
101946-152
101946-154
Ethyl-2-(1,3-benzothiazol-2-yl)-2-oxoacetate ≥95%
Ethyl-2-(1,3-benzothiazol-2-yl)-2-oxoacetate
Hazard: Irritant
Formula:
C₁₁H₉NO₃S MW: 235.26 g/mol |
MDL Number:
MFCD19443897 CAS Number: 20474-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...