Ethyl-2-(2-methyl-1,2,4-triazol-3-yl)-2-oxoacetate ≥95%

Supplier: Matrix Scientific

070740-1G 070740-5G
101946-146EA 2692.31 USD
101946-146 101946-144
Ethyl-2-(2-methyl-1,2,4-triazol-3-yl)-2-oxoacetate ≥95%
Ethyl-2-(2-methyl-1,2,4-triazol-3-yl)-2-oxoacetate

Hazard: Irritant

Formula: C₇H₉N₃O₃
MDL Number: MFCD19443900
CAS Number: 1313739-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR