1,1-Cyclobutanedimethanol ≥97%

Supplier: Matrix Scientific

070728-250MG 070728-1G 070728-500MG
101946-076EA 815.38 USD
101946-076 101946-078 101946-080
1,1-Cyclobutanedimethanol ≥97%
1,1-Cyclobutanedimethanol

Hazard: Irritant

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00463706
CAS Number: 4415-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR