9-Methylfluorene-9-carboxylic acid ≥97%
Supplier: Matrix Scientific
070709-5G
070709-1G
070709-25G
101946-006EA
1318.46
USD
101946-006
101946-004
101946-008
9-Methylfluorene-9-carboxylic acid ≥97%
9-Methylfluorene-9-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₂O₂ |
MDL Number:
MFCD00114671 CAS Number: 3300-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...