3,3-Difluorocyclobutanecarbonitrile ≥95%

Supplier: Matrix Scientific

070701-500MG 070701-250MG
101945-974EA 1221.54 USD
101945-974 101945-976
3,3-Difluorocyclobutanecarbonitrile ≥95%
3,3-Difluorocyclobutanecarbonitrile

Hazard: Irritant

Formula: C₅H₅F₂N
MDL Number: MFCD10001482
CAS Number: 86770-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR