2-Azaspiro[5.5]undecane-2,9-dicarboxylic acid-2-tert-butyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(tert-Butoxycarbonyl)-2-azaspiro[5.5]undecane-9-carboxylic acid
070697-1G
070697-250MG
070697-500MG
101945-950EA
609.5
USD
101945-950
101945-952
101945-954
2-Azaspiro[5.5]undecane-2,9-dicarboxylic acid-2-tert-butyl ester ≥95%
2-Azaspiro[5.5]undecane-2,9-dicarboxylic acid-2-tert-butyl ester
Hazard: Irritant
Formula:
C₁₆H₂₇NO₄ MW: 297.4 g/mol |
MDL Number:
MFCD18839241 CAS Number: 1251008-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...