2-Hydroxymethyloxetane ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Epoxybutanol2-OxetanemethanolOxetan-2-ylmethanol

070691-250MG
101945-926EA 238.46 USD
101945-926
2-Hydroxymethyloxetane ≥95%
2-Hydroxymethyloxetane

Hazard: Irritant

Formula: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD00142933
CAS Number: 61266-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR