6-Ethoxyimidazo[1,2-b]pyridazine ≥96%

Supplier: Matrix Scientific

070689-1G 070689-5G
101945-916EA 884.62 USD
101945-916 101945-914
6-Ethoxyimidazo[1,2-b]pyridazine ≥96%
6-Ethoxyimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₈H₉N₃O
MDL Number: MFCD18782899
CAS Number: 57470-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR