Cyclopropyldiphenylsulfonium tetrafluoroborate ≥95%
Supplier: Matrix Scientific
070688-1G
070688-5G
101945-912EA
892.31
USD
101945-912
101945-910
Cyclopropyldiphenylsulfonium tetrafluoroborate ≥95%
Cyclopropyldiphenylsulfonium tetrafluoroborate
Purity: 95%
CAS: 33462-81-6
MDL Number: MFCD00011873
Molecular Formula: C15H15BF4S
Molecular Weight: 314,16
Hazard: Irritant
Formula:
C₁₅H₁₅BF₄S MW: 314.15 g/mol |
MDL Number:
MFCD00011873 CAS Number: 33462-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...