3,6-Diazaspiro[3.3]heptane-6-carboxylic acid tert-butyl ester hemioxylate ≥95%
Supplier: Matrix Scientific
070682-500MG
070682-250MG
070682-100MG
101945-882EA
2143.08
USD
101945-882
101945-884
101945-880
3,6-Diazaspiro[3.3]heptane-6-carboxylic acid tert-butyl ester hemioxylate ≥95%
3,6-Diazaspiro[3.3]heptane-6-carboxylic acid tert-butyl ester hemioxylate
Hazard: Irritant
Formula:
C₂₂H₃₈N₄O₈ MW: 486.57 g/mol |
MDL Number:
MFCD18782996 CAS Number: 1272412-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...