2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester ≥98%

Supplier: Matrix Scientific

Synonyms: N-Boc-2,5-diaza-bicyclo[2.2.1]heptanetert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

070674-1G
101945-864EA 380 USD
101945-864
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester ≥98%
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₈N₂O₂
MW: 198.27 g/mol
MDL Number: MFCD06411671
CAS Number: 198989-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR