tert-Butyl-N-tetrahydrothiopyran-4-ylcarbamate ≥97%

Supplier: Matrix Scientific

070670-5G 070670-1G
101945-840EA 2416.92 USD
101945-840 101945-838
tert-Butyl-N-tetrahydrothiopyran-4-ylcarbamate ≥97%
tert-Butyl-N-tetrahydrothiopyran-4-ylcarbamate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂S
MW: 217.33 g/mol
MDL Number: MFCD16659876
CAS Number: 595597-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR