5-Bromo-2-chloro-3,4-pyridinediamine ≥96%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-chloropyridine-3,4-diamine
070664-1G
070664-5G
070664-500MG
101945-808EA
640
USD
101945-808
101945-812
101945-810
5-Bromo-2-chloro-3,4-pyridinediamine ≥96%
5-Bromo-2-chloro-3,4-pyridinediamine
Hazard: Irritant
Formula:
C₅H₅BrClN₃ MW: 222.48 g/mol |
MDL Number:
MFCD02266941 CAS Number: 163452-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...